Assay Details
Gene Symbol: LOC100356924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002721532.3 | XP_002721578.1 | 492 | 105 | ||
XM_017337603.1 | XP_017193092.1 | 559 | 105 |

More Information
Set Membership: |
![]() ![]() |
