Assay Details
Gene Symbol: LOC100356924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002721532.3 | XP_002721578.1 | 542 | 81 | ||
XM_008275417.2 | XP_008273639.1 | 191 | 81 | |||
XM_017337603.1 | XP_017193092.1 | 609 | 81 |

More Information
Set Membership: |
![]() ![]() |
