Assay Details
Gene Symbol: TMPRSS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008256457.2 | XP_008254679.1 | 3 - 4 | 719 | 136 | |
XM_017341856.1 | XP_017197345.1 | 3 - 4 | 719 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
