Assay Details
Gene Symbol: AKNAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264772.2 | XP_008262994.1 | 11 - 12 | 1954 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AKNAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264772.2 | XP_008262994.1 | 11 - 12 | 1954 | 100 |
Set Membership: |
![]() ![]() ![]() ![]() |