Assay Details
Gene Symbol: FBXL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008270409.2 | XP_008268631.1 | 1 - 2 | 221 | 80 | |
XM_017348521.1 | XP_017204010.1 | 1 - 2 | 221 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008270409.2, XM_017348521.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
