Assay Details
Gene Symbol: TENT5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017349678.1 | XP_017205167.1 | 3 - 4 | 301 | 112 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017349678.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
