Assay Details
Gene Symbol: BLM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008275317.2 | XP_008273539.1 | 1275 | 92 | ||
XM_008275320.2 | XP_008273542.1 | 1275 | 92 |

More Information
Set Membership: |
![]() ![]() |
