Assay Details
Gene Symbol: AKAP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008272936.2 | XP_008271158.1 | 5 - 6 | 637 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
