Assay Details
Gene Symbol: GPRC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008257774.2 | XP_008255996.1 | 3 - 4 | 1477 | 68 | |
XM_008257775.1 | XP_008255997.1 | 3 - 4 | 1421 | 68 | ||
XM_008257777.2 | XP_008255999.1 | 3 - 4 | 1418 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
