Assay Details
Gene Symbol: CPLANE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002722334.3 | XP_002722380.1 | 150 | 94 | ||
XM_017338275.1 | XP_017193764.1 | 153 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002722334.3, XM_017338275.1 transcripts.
Set Membership: |
![]() ![]() |
