Assay Details
Gene Symbol: CPLANE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002722334.3 | XP_002722380.1 | 271 | 107 | ||
XM_017338275.1 | XP_017193764.1 | 274 | 107 |

More Information
Set Membership: |
![]() ![]() |
