Assay Details
Gene Symbol: PMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712339.3 | XP_002712385.1 | 3 - 4 | 382 | 71 | |
XM_008258829.2 | XP_008257051.1 | 3 - 4 | 362 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
