Assay Details
Gene Symbol: MFSD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002708223.3 | XP_002708269.1 | 11 - 12 | 1661 | 69 | |
XM_008256805.2 | XP_008255027.1 | 10 - 11 | 1589 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
