Assay Details
Gene Symbol: EEF1AKNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002715894.3 | XP_002715940.1 | 3 - 4 | 2604 | 97 | |
XM_008264068.2 | XP_008262290.1 | 4 - 5 | 2390 | 97 | ||
XM_008264069.2 | XP_008262291.1 | 3 - 4 | 1097 | 97 | ||
XM_008264070.2 | XP_008262292.1 | 3 - 4 | 1075 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
