Assay Details
Gene Symbol: EEF1AKNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002715894.3 | XP_002715940.1 | 7 - 8 | 3314 | 64 | |
XM_008264068.2 | XP_008262290.1 | 8 - 9 | 3100 | 64 | ||
XM_008264069.2 | XP_008262291.1 | 7 - 8 | 1807 | 64 | ||
XM_008264070.2 | XP_008262292.1 | 7 - 8 | 1785 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
