Assay Details
Gene Symbol: MAP3K21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717355.3 | XP_002717401.1 | 8 - 9 | 2205 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MAP3K21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717355.3 | XP_002717401.1 | 8 - 9 | 2205 | 104 |
Set Membership: |
![]() ![]() ![]() |