Assay Details
Gene Symbol: ENY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710737.3 | XP_002710783.1 | 4 - 5 | 368 | 67 | |
XM_008255833.1 | XP_008254055.1 | 4 - 5 | 306 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
