Assay Details
Gene Symbol: YWHAE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718890.3 | XP_002718936.1 | 2 - 3 | 376 | 106 | |
XM_008270937.2 | XP_008269159.1 | 2 - 3 | 378 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
