Assay Details
Gene Symbol: SERPINB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720957.3 | XP_002721003.1 | 189 | 55 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002720957.3 transcripts.
Set Membership: |
![]() ![]() |
