Assay Details
Gene Symbol: ZCCHC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002707981.3 | XP_002708027.1 | 5 - 6 | 1048 | 65 | |
XM_017337500.1 | XP_017192989.1 | 6 - 7 | 1276 | 65 | ||
XM_017350928.1 | XP_017206417.1 | 5 - 6 | 1086 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
