Assay Details
Gene Symbol: ECHDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717616.3 | XP_002717662.1 | 4 - 5 | 706 | 96 | |
XM_017347674.1 | XP_017203163.1 | 5 - 6 | 768 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
