Assay Details
Gene Symbol: LOC100350827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262483.2 | XP_008260705.1 | 10 - 11 | 1317 | 92 | |
XM_008262484.2 | XP_008260706.1 | 8 - 9 | 1211 | 92 | ||
XM_017344959.1 | XP_017200448.1 | 11 - 12 | 1421 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
