Assay Details
Gene Symbol: LOC100350827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262483.2 | XP_008260705.1 | 14 - 15 | 1876 | 76 | |
XM_008262484.2 | XP_008260706.1 | 12 - 13 | 1770 | 76 | ||
XM_017344959.1 | XP_017200448.1 | 15 - 16 | 1980 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
