Assay Details
Gene Symbol: LOC100350827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262483.2 | XP_008260705.1 | 2 - 3 | 222 | 85 | |
XM_017344959.1 | XP_017200448.1 | 3 - 4 | 326 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008262483.2, XM_017344959.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
