Assay Details
Gene Symbol: LOC100350827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262483.2 | XP_008260705.1 | 9 - 10 | 1184 | 98 | |
XM_008262484.2 | XP_008260706.1 | 7 - 8 | 1078 | 98 | ||
XM_017344959.1 | XP_017200448.1 | 10 - 11 | 1288 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
