Assay Details
Gene Symbol: KIRREL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264256.2 | XP_008262478.1 | 7 - 8 | 1025 | 93 | |
XM_008264257.2 | XP_008262479.1 | 7 - 8 | 1025 | 93 | ||
XM_008264258.2 | XP_008262480.1 | 6 - 7 | 875 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
