Assay Details
Gene Symbol: GRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002722658.3 | XP_002722704.1 | 402 | 67 | ||
XM_008250354.1 | XP_008248576.1 | 402 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
