Assay Details
Gene Symbol: GNA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008256681.2 | XP_008254903.1 | 4 - 5 | 659 | 64 | |
XM_008256685.2 | XP_008254907.1 | 3 - 4 | 424 | 64 | ||
XM_017341945.1 | XP_017197434.1 | 3 - 4 | 1346 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
