Assay Details
Gene Symbol: LMNTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008259504.2 | XP_008257726.1 | 5 - 6 | 992 | 77 | |
XM_017343321.1 | XP_017198810.1 | 5 - 6 | 993 | 77 | ||
XM_017343322.1 | XP_017198811.1 | 5 - 6 | 992 | 77 | ||
XM_017343323.1 | XP_017198812.1 | 5 - 6 | 992 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
