Assay Details
Gene Symbol: FAM163A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002715008.3 | XP_002715054.1 | 2 - 3 | 272 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002715008.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
