Assay Details
Gene Symbol: ALKAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002723190.3 | XP_002723236.1 | 381 | 99 | ||
XM_008251378.2 | XP_008249600.1 | 610 | 99 | |||
XM_017339209.1 | XP_017194698.1 | 382 | 99 | |||
XM_017339210.1 | XP_017194699.1 | 381 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
