Assay Details
Gene Symbol: DYNC2I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008251000.2 | XP_008249222.1 | 1384 | 83 | ||
XM_008251001.2 | XP_008249223.1 | 1326 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
