Assay Details
Gene Symbol: EMSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002708680.3 | XP_002708726.1 | 8 - 9 | 1246 | 65 | |
XM_002708681.3 | XP_002708727.1 | 9 - 10 | 1289 | 65 | ||
XM_002708682.3 | XP_002708728.1 | 8 - 9 | 1135 | 65 | ||
XM_002708683.3 | XP_002708729.1 | 9 - 10 | 1288 | 65 | ||
XM_008263753.2 | XP_008261975.1 | 7 - 8 | 1171 | 65 | ||
XM_008263756.2 | XP_008261978.1 | 8 - 9 | 1135 | 65 | ||
XM_008263759.2 | XP_008261981.1 | 9 - 10 | 1289 | 65 | ||
XM_008263768.2 | XP_008261990.1 | 7 - 8 | 1093 | 65 | ||
XM_008263782.2 | XP_008262004.1 | 9 - 10 | 1288 | 65 | ||
XM_008263786.2 | XP_008262008.1 | 9 - 10 | 1289 | 65 | ||
XM_008263790.2 | XP_008262012.1 | 8 - 9 | 1246 | 65 | ||
XM_017344550.1 | XP_017200039.1 | 9 - 10 | 1286 | 65 | ||
XM_017344561.1 | XP_017200050.1 | 8 - 9 | 1243 | 65 | ||
XM_017344592.1 | XP_017200081.1 | 9 - 10 | 1288 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
