Assay Details
Gene Symbol: ELP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709032.3 | XP_002709078.1 | 3 - 4 | 420 | 71 | |
XM_008269353.2 | XP_008267575.1 | 3 - 4 | 420 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
