Assay Details
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008271854.2 | XP_008270076.1 | 1 - 2 | 145 | 89 | |
XM_017346737.1 | XP_017202226.1 | 1 - 2 | 145 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008271854.2, XM_017346737.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
