Assay Details
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008271854.2 | XP_008270076.1 | 3 - 4 | 430 | 86 | |
XM_008271859.2 | XP_008270081.1 | 4 - 5 | 812 | 86 | ||
XM_017346737.1 | XP_017202226.1 | 3 - 4 | 430 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
