Assay Details
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008271854.2 | XP_008270076.1 | 6 - 7 | 747 | 77 | |
XM_008271859.2 | XP_008270081.1 | 7 - 8 | 1129 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
