Assay Details
Gene Symbol: LOC100345427 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002714462.3 | XP_002714508.2 | 5 - 6 | 962 | 115 | |
XM_017345162.1 | XP_017200651.1 | 5 - 6 | 964 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
