Assay Details
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008272223.2 | XP_008270445.1 | 1 - 2 | 104 | 55 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008272223.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
