Assay Details
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008272222.2 | XP_008270444.1 | 3 - 4 | 277 | 75 | |
XM_008272223.2 | XP_008270445.1 | 2 - 3 | 293 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
