Assay Details
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008272222.2 | XP_008270444.1 | 4 - 5 | 508 | 94 | |
XM_008272223.2 | XP_008270445.1 | 3 - 4 | 524 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
