Assay Details
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002714289.3 | XP_002714335.1 | 8 - 9 | 1360 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002714289.3 | XP_002714335.1 | 8 - 9 | 1360 | 58 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |