Assay Details
Gene Symbol: LOC100345657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008249573.2 | XP_008247795.1 | 2 - 3 | 238 | 104 | |
XM_008249579.2 | XP_008247801.1 | 2 - 3 | 169 | 104 | ||
XM_008249585.2 | XP_008247807.1 | 2 - 3 | 238 | 104 | ||
XM_008249590.2 | XP_008247812.1 | 2 - 3 | 238 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008249573.2, XM_008249579.2, XM_008249585.2, XM_008249590.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
