Assay Details
Gene Symbol: UBE2L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709162.3 | XP_002709208.1 | 2 - 3 | 236 | 80 | |
XM_017346623.1 | XP_017202112.1 | 2 - 3 | 418 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
