Assay Details
Gene Symbol: BBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709022.3 | XP_002709068.1 | 6 - 7 | 1292 | 88 | |
XM_008269060.2 | XP_008267282.1 | 6 - 7 | 1294 | 88 | ||
XM_008269063.2 | XP_008267285.1 | 6 - 7 | 1237 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
