Assay Details
Gene Symbol: LAMTOR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002724248.3 | XP_002724294.2 | 127 | 99 | ||
XM_008253581.2 | XP_008251803.1 | 128 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002724248.3, XM_008253581.2 transcripts.
Set Membership: |
![]() ![]() |
