Assay Details
Gene Symbol: PLA2G12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718428.3 | XP_002718474.1 | 2 - 3 | 443 | 79 | |
XM_008269974.2 | XP_008268196.1 | 2 - 3 | 445 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
