Assay Details
Gene Symbol: TRAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718919.3 | XP_002718965.1 | 1 - 2 | 140 | 80 | |
XM_017348951.1 | XP_017204440.1 | 1 - 2 | 238 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
