Assay Details
Gene Symbol: MYZAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717707.3 | XP_002717753.1 | 12 - 13 | 1428 | 69 | |
XM_008268955.1 | XP_008267177.1 | 13 - 14 | 1507 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
