Assay Details
Gene Symbol: MYZAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717707.3 | XP_002717753.1 | 3 - 4 | 445 | 83 | |
XM_008268955.1 | XP_008267177.1 | 4 - 5 | 524 | 83 | ||
XM_008268956.2 | XP_008267178.1 | 4 - 5 | 526 | 83 | ||
XM_008268957.2 | XP_008267179.1 | 3 - 4 | 446 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
